Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551907 Supplier Invitrogen™ Supplier No. PA551907
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLVPIKPAPL PLSPGKNSFG ILSSKGNILQ IQGSQLSASY PGGQLVFAIQ NPTMINKGTR SNANIQYQAV PQIQASNSQT IQVQPNLTNQ IQIIPGTNQA IITPSPSSHK PVPIKPAPIQ KSST Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

This gene encodes a member of the Sp subfamily of Sp/XKLF transcription factors. Sp family proteins are sequence-specific DNA-binding proteins characterized by an amino-terminal trans-activation domain and three carboxy-terminal zinc finger motifs. This protein contains the least conserved DNA-binding domain within the Sp subfamily of proteins, and its DNA sequence specificity differs from the other Sp proteins. It localizes primarily within subnuclear foci associated with the nuclear matrix, and can activate or in some cases repress expression from different promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SP2
Gene Accession No. Q02086
Gene Alias 4930480I16Rik; Kiaa0048; mKIAA0048; Sp2; Sp2 transcription factor; Trans-acting transcription factor 2; transcription factor Sp2
Gene Symbols SP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SP2. Recombinant protein control fragment (Product #RP-100740).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.