Learn More
Invitrogen™ SP3 Monoclonal Antibody (4E5)

Description
Peptide Sequence: TAGINADGHL INTGQAMDSS DNSERTGERV SPDINETNTD TDLFVPTSSS SQLPVTIDST GILQQNTNSL TTSSGQVHSS DLQGNYIQSP VSEETQAQNI QVSTAQPVVQ HLQLQESQQP TSQAQIVQGI TPQTIHGVQA SGQN*.
Specifications
Specifications
| Antigen | SP3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SP3 |
| Gene Accession No. | O70494, Q02447 |
| Gene Alias | D130027J01Rik; DKFZp686O1631; GC-binding transcription factor Sp3; SP3; Sp3 transcription factor; specificity protein 3; SPR1; SPR-1; SPR2; SPR-2; trans-acting transcription factor 3; transcription factor Sp3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.