Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Spadin
GREENER_CHOICE

Catalog No. 55941
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55941 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 55941 Supplier Tocris Bioscience™ Supplier No. 5594/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent K2P2.1 (TREK-1) channel blocker

Spadin is a potent TREK-1 channel blocker (IC50 = 71 nM). Enhances dorsal raphe nucleus 5-HT neurotransmission in mice. Induces hippocampal CREB activation and neurogenesis in adult mice. Exhibits antidepressant effects in mouse models of depression. Brain penetrant.

Specifications

Purity 0.95
Description Spadin
Molecular Weight (g/mol) 2012.34
Product Type Spadin
Quantity 1 mg
Sequence Tyr-Ala-Pro-Leu-Pro-Arg-Trp-Ser-Gly-Pro-Ile-Gly-Val-Ser-Trp-Gly-Leu-Arg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.