Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100185 Supplier Invitrogen™ Supplier No. PA5100185
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SPAG4L.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG4L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SUN5
Gene Accession No. Q8TC36, Q9DA32
Gene Alias 1700021O15Rik; dJ726C3.1; Sad1 and UNC84 domain containing 5; sad1 and UNC84 domain-containing protein 5; SPAG4L; SPAG4L-2; Sperm-associated antigen 4-like protein; SRG4; SUN domain-containing protein 5; SUN5; testis and spermatogenesis related gene 4; testis and spermatogenesis-related gene 4 protein; TSARG4
Gene Symbols SUN5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SUN5(Accession Q8TC36), corresponding to amino acid residues A266-F316.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140732, 76407
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.