Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-7184229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110877 Supplier Invitrogen™ Supplier No. PA5110877
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTVENLTAKL ASTIADNQEQ DLEKTRQYSQ KLGLLTEQLQ SLTLFLQTKL KEKTEQETLL LSTACPPTQE HPLPNDRTFL GSILTAVADE EPESTPVPLL GSDKSAFTRV ASMVSL Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 60%.

SPAG5 (sperm associated antigen 5), also known as MAP126, hMAP126, DEEPEST or Astrin, is a 1,193 amino acid protein that is expressed at high levels in testis and at lower levels in liver, placenta, pancreas, colon and thymus. Localized specifically to spindle poles during prophase and throughout the spindle during metaphase and anaphase, SPAG5 is essential for proper spindle formation and is thought to be involved in the dynamic and functional regulation of mitotic spindles. Additionally, SPAG5 plays a role in sister chromatid cohesion and subsequent chromatid separation and, via its association with chromatids, is an important regulator of chromosome integrity. SPAG5 has a long stalk and a globular head domain through which if forms a homodimer that is characterized by aster-like structures. Upon DNA damage, SPAG5 may be phosphorylated by ATR or ATM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPAG5
Gene Accession No. Q96R06
Gene Alias AI874642; Astrin; D11Bhm180e; Deepest; hMAP126; MAP126; Mastrin; microtubule-associated protein; mitotic spindle associated protein p126; mitotic spindle coiled-coil related protein; mitotic spindle-associated protein p126; S17; Spag5; sperm associated antigen 5; sperm-associated antigen 5
Gene Symbols SPAG5
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SPAG5. Recombinant protein control fragment (Product #RP-93178).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10615
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.