Learn More
Invitrogen™ SPANXB1 Monoclonal Antibody (OTI1E11)

Description
SPANXB1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SPANXB1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SPANXB1 |
| Gene Accession No. | Q9NS25 |
| Gene Alias | B1; cancer/testis antigen 11.2; cancer/testis antigen family 11, member 2; CT11.2; Nuclear-associated protein SPAN-Xb; nuclear-associated protein SPAN-Xf; SPANX family member B/F; SPANX family member B1; SPANX family member F1; SPANX family, member B1; SPANX family, member B2; SPANX family, member F1; SPANX family, member F2; SPANXB; SPANX-B; SPANXB1; SPANXB2; SPANXF1; SPANXF2; sperm protein associated with the nucleus on the X chromosome B/F; sperm protein associated with the nucleus on the X chromosome B1; sperm protein associated with the nucleus, X chromosome, family member B1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.