Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sparfloxacin, 98%

Catalog No. AAJ6635803
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6635803 1 g
AAJ6635809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6635803 Supplier Thermo Scientific Chemicals Supplier No. J6635803
Add to Cart

CAS: 110871-86-8 | C19H22F2N4O3 | 392.41 g/mol

Targets a wide range of Gram-positive, Gram-negative, and mycoplasma species

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110871-86-8
Molecular Formula C19H22F2N4O3
Molecular Weight (g/mol) 392.41
MDL Number MFCD00869619
InChI Key DZZWHBIBMUVIIW-DTORHVGOSA-N
Synonym sparfloxacin, zagam, esparfloxacino, sparfloxacine, sparfloxacinum, spara, sparfloxacine inn-french, sparfloxacinum inn-latin, spfx, esparfloxacino inn-spanish
PubChem CID 60464
ChEBI CHEBI:9212
IUPAC Name 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O

Specifications

Melting Point 265°C
Boiling Point 640°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 1 g
Beilstein 9170271
Solubility Information Soluble in DMSO at 9mg/ml. Sparingly soluble in water
Formula Weight 392.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sparfloxacin
Show More Show Less
RTECSNumber VB1986500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.