Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Spark Timer
SDPSCI_ED

Catalog No. S81504 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81504 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81504 Supplier United Scientific™ Supplier No. SPARK1
Add to Cart
Add to Cart

Spark Timer is used for measuring velocity and acceleration of falling or moving objects.

  • Spark timer is used for measuring velocity and acceleration of falling or moving objects.
  • When the tape is pulled through the timer, a spark is created which burns tiny dots in the tape at the same, precise interval.
  • The faster the tape is pulled through the timer, the larger the distance between the dots.
  • This allows students to calculate the acceleration.
  • Spark tape, carbon disks, weight and clamp are included.
  • Includes activity guide.

Specifications

Grade 5+
Electrical Requirements 110 V, US Plug
Format Kit
Product Type Spark Timer
Includes Activity guide
Material Plastic
Languages English
For Use With (Application) Measuring velocity and acceleration.
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.