Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA22 Monoclonal Antibody (OTI1E2)
GREENER_CHOICE

Catalog No. PIMA526991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526991 Supplier Invitrogen™ Supplier No. MA526991
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA22 Monoclonal Antibody for Western Blot, IHC (P)

SPATA22 is highly expressed in adult testis. The specific function of SPATA22 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPATA22
Gene Accession No. Q8NHS9
Gene Alias Gm882; NYDSP20; NYD-SP20; Repro42; RGD1565378; Sgn; Spata22; spermatogenesis associated 22; spermatogenesis-associated protein 22; testicular tissue protein Li 186; testis development protein NYD-SP20; Unknown (protein for MGC:134099)
Gene Symbols SPATA22
Host Species Mouse
Immunogen Full length human recombinant protein of SPATA22 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.