Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA2L Monoclonal Antibody (OTI3H8), TrueMAB™, OriGene
SDP

Catalog No. 501679208
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9208 Supplier OriGene Supplier No. TA802363
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.

Specifications

Antigen SPATA2L
Applications Western Blot
Classification Monoclonal
Clone OTI3H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SPATA2L
Gene Accession No. Q8IUW3
Gene Alias C16orf76, tamo
Gene Symbols SPATA2L
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 179-424 of human SPATA2L produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124044
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.