Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Spatula Round End and Tapered Flat End, Stainless Steel

Catalog No. 14127255
Change view
Click to view available options
Length (Metric):
19.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
14-127-255 19.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-255 Supplier Heathrow Scientific Supplier No. HS15907
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spatula transfers, lifts, and handles powders and granular materials.

  • Made from nickel-plated, stainless steel wire
  • Mirror finish with round flat and tapered flat ends
  • Autoclavable

Specifications

Description Spatula Stainless Steel
Coating Nickel-Plated
Color Silver
Disposable Reusable
Product Type Spatula
Finish Mirror
For Use With (Application) Powders, Granular Material
Handle Color Silver
Length (English) 7.7 in.
Blade Material Stainless Steel
Handle Material Stainless Steel
Material Nickel/Stainless Steel
Handle Type Round Flat
Length (Metric) 19.5 cm
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round End, Tapered End
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.