Learn More
Perkin Elmer US LLC SPB 100-42, 230 V
Supplier:  Perkin Elmer US LLC N9300807
Encompass_Preferred
The SPB series of block digestion systems include graphite blocks that are PTFE-coated to resist aggressive corrosive attack for guaranteed long life in harsh laboratory environments.
Catalog No. 50-210-4243
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.