Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC™ Absorbents HAZWIK™ Quick Response Kits

Catalog No. 19129878 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Application):
For Bodily Fluids
For Chemical Spills
Capacity (English):
0.33 gal.
0.49 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Capacity (English)
19-129-878 For Chemical Spills 0.33 gal.
19-129-879 For Bodily Fluids 0.49 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-129-878 Supplier Brady™ SPC™ Absorbents Supplier No. SKHAZQRKC5
Add to Cart
Add to Cart

The HAZWIK™ Quick Response Kit provides fast and effective cleanup for emergency spills in a convenient, grab-and-go-style bag.

  • It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
  • Compact, single-use kit that is ideal for use on small spills in the lab.
  • Quickly cleans up hazardous chemical spills.
  • Visual instructions on the package.
  • Contains PPE, tools, absorbent pads, acid and base neutralizers, wipes and disposable materials.
  • Five individual kits per case.

Specifications

Color Green
Description Ideal for use on small spills in the lab
Capacity (English) 0.33 gal.
Material Polyethylene
Type Emergency Spill Kit
For Use With (Application) For Chemical Spills
Includes 7.5 x 10 in. Pads, 6; 4.5 oz. Acid Neutralizer, 1; 4.5 oz. Base Neutralizer, 1; Pair of Nitrile Gloves, 1; Scoop and Scraper, 1; Disposal Bag, 1; Cable Tie, 1; Surface Sanitizing Wipe, 1
Width (English) 9 in.
Height (English) 13.13 in.
Depth (English) 19 in.
Weight (English) 0.7 lb.
Weight (Metric) 1.54 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.