Learn More
Brady™ SPC™ MAXX Enhanced Heavy Weight Oil Rolls
Catalog No. 17111175 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
70 gal./Bale
72 gal./Bale
Dimensions (L x W):
144 ft. x 19 in.
144 ft. x 38 in
Packaging Quantity:
1 Ea.
2/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W) Packaging Quantity
17-111-175 70 gal./Bale 144 ft. x 38 in 1 Ea.
17-111-189 72 gal./Bale 144 ft. x 19 in. 2/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-175 Supplier Brady™ Supplier No. SPC150
Add to Cart
Add to Cart
For use with oil and petroleum based fluids
  • Single-ply construction with MAXX technology
  • Hydrophobic nature for outdoor applications
  • Floats indefinitely even when saturated with oil
  • Dimpled and perforated
  • Heavier and bonded to reduce linting
  • Available in various sizes
  • Specifications
    Capacity (English) 70 gal./Bale
    Material Polypropylene
    Type Oil Absorbent Roll
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds
    Packaging Quantity 1 Ea.
    Dimensions (L x W) 144 ft. x 38 in