Learn More
Newcomer Supply Specimen Chuck w/Colored O-ring & Orientation Mark, Round, 30 mm, Orange
SDP
Supplier:  Newcomer Supply 6865E
Encompass
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.
Catalog No. NC1841640
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.