Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 3-oxo-1-(pyridin-2-yldisulfanyl)-PEG24 oate | 00-00-0 | MFCD13185007 | >98.0% | C63H113N3O29S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14135710MG
Encompass_Preferred
SPDP-PEG24-NHS ester is a heterobifunctional PEG linker with an N-hydroxysuccinimide (NHS) ester at one end and a pyridyldithiol (SPDP) group at the other. It enables selective amine-to-thiol conjugation for bioconjugation and PROTAC assembly, and is supplied in small research quantities with high reported purity.
  • Enables amine-to-thiol conjugation through NHS and pyridyldithiol chemistries.
  • Provides a hydrophilic PEG24 spacer to improve solubility and flexibility.
  • Suitable for use in PROTAC synthesis and bioconjugation workflows.
  • Supplied in small research quantities with high reported purity (>98%).
  • Molecular weight approximately 1440.7 g/mol to aid stoichiometry planning.
Catalog No. 50-002-60703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.