Learn More
MilliporeSigma™ Supelco™ SPE Tube Adapter PTFE (with female luer port)
Catalog No. 111008906 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
24/Pk.
Catalog No. Packaging Quantity
11-100-8906 24/Pk.
1 options
Catalog No. 11-100-8906 Supplier MilliporeSigma™ Supelco™ Supplier No. 504335
Add to Cart
Add to Cart
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 24/Pk.
For Use With 6 mL Glass SPE Tubes
Material PTFE