Learn More
MilliporeSigma™ Supelco™ SPE Tube Adapter
Catalog No. 111020889 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
12/Pk.
Catalog No. Packaging Quantity
11-102-0889 12/Pk.
1 options
Catalog No. 11-102-0889 Supplier MilliporeSigma™ Supelco™ Supplier No. 57020U
Add to Cart
Add to Cart
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 12/Pk.
For Use With 1, 3, 6 mL Tubes