Learn More
ORS Nasco SPEC ASTRO PAT/CL XTREME 10EA
Supplier:  ORS Nasco 763S2530C
Encompass_Preferred
Honeywell part# S2530C. Honeywell Safety Products Uvex Astro OTG 3001 Safety Glasses Frame Color: Red; White; Blue Lens Type: Anti-fog Lens Tint: Clear Frame Material: Nylon Lens Coating: Uvextreme Anti-fog Lens Material: Polycarbonate UV Protection: 99.90%
Catalog No. 50-255-1584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.