Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPECC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571632
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571632 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571632 Supplier Invitrogen™ Supplier No. PA571632
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPECC1L Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein that is leucine-rich and is thought to play a role in regulating the interaction of the cytoskeleton with a variety of cellular processes. Mutations in this gene are associated with the French-Canadian type of Leigh syndrome. Transcripts ranging in size from 4. 8 to 7. 0 kb which result from alternative polyadenylation have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPECC1L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SPECC1L
Gene Accession No. Q2KN98, Q69YQ0
Gene Alias 4930470P14Rik; 4932439K10Rik; 9530057A13Rik; AI317249; cytokinesis and spindle organization A; cytospin A; cytospin-A; CYTSA; GBBB2; Kiaa0376; mKIAA0376; OBLFC1; Renal carcinoma antigen NY-REN-22; RGD1309570; Specc1l; SPECC1-like; SPECC1-like protein; sperm antigen with calponin homology and coiled-coil domains 1 like; sperm antigen with calponin homology and coiled-coil domains 1-like
Gene Symbols SPECC1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 108-136 amino acids from the N-terminal region of human CYTSA.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23384, 74392
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.