Learn More
Market Lab Specimen Collection Container, 4oz, Sterile, 100pk
Supplier:  Market Lab 16431GN
Encompass_Preferred
The ML16431-GN is a sterile specimen collection container made by MarketLab. It has a 50mL capacity and is made from clear polyethylene plastic. The container has a leakproof screw cap and graduated markings up to 50mL. It can be used to collect, transport, and store urine, sputum, and other biological specimens. The container is latex-free, non-pyrogenic, and for single use. It is sold in cases of 500.
Catalog No. NC1926374
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.