Learn More
Perkin Elmer US LLC Special Quartz Macro Cuvette with Cover - 50 mm, Qty. 1
Supplier:  Perkin Elmer US LLC N4103014
Encompass_Preferred
These durable, fire-fused cells are standard rectangular spectrophotometer cuvettes with two optically-polished windows. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size UV Quartz 50 mm 52.5 mm 12.5 mm 17.5 mL Macro
Catalog No. 50-209-3280
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.