Learn More
Eisco™ Specific Gravity Cubes
SCI_ED
Catalog No. S99390 Shop All Eisco Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
S99390 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. S99390 Supplier Eisco™ Supplier No. PH0108
Add to Cart
Add to Cart
This item is not returnable. View return policy
Investigate a range of specific gravities.
  • 0.79 in. (20mm) cubes with hooks
  • Set includes brass, iron, aluminum and zinc cubes
Specifications
Quantity 4/Pk.
For Use With (Application) Measure the specific gravity
Product Type Specific Gravity Cube
Includes Set of four cube made of Brass, Iron, Aluminum, Zinc with hooks
Material Brass, Iron, Aluminum, Zinc
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov