Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Specific Gravity Cylinders
SCI_ED

Catalog No. S24863 Shop All Eisco Products
Change view
Click to view available options
Grade:
8 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S24863 8 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24863 Supplier Eisco™ Supplier No. PH0112ACB
Add to Cart
Add to Cart

Help students understand density with these Specific Gravity Cylinders.

  • With hook
  • Set of five metals including Aluminum, Copper, Steel, Tin and Zinc
  • Instruction manual with details on density, specific gravity, specific heat
  • Contains three inquiry-based student activities with capture sheets
  • Each cylinder with 0.5 in. dia. and mass and different length
  • Hooks used to measure mass using spring scale

Specifications

Grade 8 to Undergraduate
Includes Set of 5 Cylinders of Zinc, Copper, Aluminum, Tin, and Steel, Complete Instruction Manual with 3 Inquiry-based Student Activities with Capture Sheets
Diameter (English) Outer 0.5 in.
Diameter (Metric) Outer 12.7 mm
Product Type Set of Specific Gravity Cylinders with Hooks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.