Learn More
Cenmed™ Specimen Bag with Indicators
SDP
Catalog No. 21520194
Change view
Click to view available options
Includes:
Indicator
Indicator, Absorbent Pad
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-520-194 Indicator
21-520-195 Indicator, Absorbent Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-520-194 Supplier Cenmed™ Supplier No. 181609B
Only null left
Add to Cart
Add to Cart
Transport specimens with Cenmed™ bags. Each bag is printed with an orange biohazard symbol and features a document pouch.
  • Bags each printed with orange biohazard symbol
  • Checkmark boxes on each bag to indicate special handling instructions
  • Document compartment attached
  • Zipper closure allows repeated access to specimen
  • Fisher catalog number 21520195: Each bag additionally contains contains a liquid absorbant pad that easily absorbs aqueous solutions or bodily fluids providing safe transportation of specimens
Specifications
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Includes Indicator
No. per Case 1000/CS.
Dimensions (L x W) 9 x 6 in.
Length (English) 9 in.
Width (English) 6 in.