Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Specimen Chucks for CRYOSTAR Cryostats, 30 mm
SDP

Catalog No. 22445744 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Purple
Red
Product Type:
Circular Chuck
Waffle Chuck
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22-445-744 Orange Circular Chuck
22-445-510 Blue Circular Chuck
22-445-646 Green Circular Chuck
22-445-325 Purple Circular Chuck
22-445-326 Red Circular Chuck
22-445-882 Blue Waffle Chuck
22-445-628 Green Waffle Chuck
22-445-304 Orange Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-445-744 Supplier Mopec™ Supplier No. SP123OR
Add to Cart
Add to Cart

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) CryoStar NX50 and NX70
Color Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.