Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Specimen Chucks for CRYOSTAR Cryostats, 40 mm
SDP

Catalog No. 22445556 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22-445-556 White Circular Chuck
22-445-306 Blue Circular Chuck
22-445-477 Green Circular Chuck
22-445-497 Orange Circular Chuck
22-445-475 Pink Circular Chuck
22-445-626 Purple Circular Chuck
22-445-327 Red Circular Chuck
22-445-575 Yellow Circular Chuck
22-445-333 Orange Waffle Chuck
22-445-658 Pink Waffle Chuck
22-445-495 Purple Waffle Chuck
22-445-313 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 22-445-556 Supplier Mopec™ Supplier No. SP124WH
Add to Cart
Add to Cart

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) CryoStar NX50 and NX70
Material Aluminum
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.