Learn More
Description
- Round, aluminum specimen chucks
Note:
Photo is representative only. Actual color may vary.
Specifications
Specifications
| Product Type | Circular Chuck |
| For Use With (Equipment) | Leica CM1950 Cryostat |
| Color | Purple |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
