Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Specimen Chucks for Leica CM1950 Cryostat, 30 mm
SDP

Catalog No. 22445811 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22-445-811 Blue Waffle Chuck
22-445-507 Blue Circular Chuck
22-445-753 Green Circular Chuck
22-445-420 Orange Circular Chuck
22-445-754 Pink Circular Chuck
22-445-422 Purple Circular Chuck
22-445-647 Red Circular Chuck
22-445-580 White Circular Chuck
22-445-679 Yellow Circular Chuck
22-445-402 Purple Waffle Chuck
22-445-812 Red Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-445-811 Supplier Mopec™ Supplier No. SP113WBL
Add to Cart
Add to Cart

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Waffle Chuck
For Use With (Equipment) Leica CM1950 Cryostat
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.