Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Specimen Chucks for QS and Microm Cryostats, 30 mm
SDP

Catalog No. 22445458 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22-445-458 Purple Waffle Chuck
22-445-694 Blue Circular Chuck
22-445-307 Green Circular Chuck
22-445-496 Orange Circular Chuck
22-445-574 Pink Circular Chuck
22-445-720 Purple Circular Chuck
22-445-504 White Circular Chuck
22-445-834 Yellow Circular Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-445-458 Supplier Mopec™ Supplier No. SP133WPR
Add to Cart
Add to Cart

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Waffle Chuck
For Use With (Equipment) QS and Microm Cryostats
Color Purple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.