Learn More
Marketlab Specimen Drop Boxes
Catalog No. 22505044
Change view
Click to view available options
Product Type:
Drop Box
Catalog No. Product Type
22-505-044 Drop Box
1 options
Catalog No. 22-505-044 Supplier Marketlab Supplier No. 0114
Add to Cart
Add to Cart
Plastic design with 14 W x 7.25 D x 11 H in. dimensions. Supports secure specimen collection and storage.
  • Key Locked Courier Specimen Drop Boxes
  • After-hours specimen boxes attach to top, side, or bottom of door, depending on model
  • Foam-lined interiors protect specimens
  • Imprinted with biohazard symbol and set of 2 keys included
  • Specimen Drop Boxes allow after-hours specimen drop-off
  • We offer two different lock box materials (steel and plastic) as well as over-the-door and on-the-side placement options
  • Deliver and guard specimens wisely with this solution
Specifications
Product Type Drop Box