Learn More
BINDER™ Specimen Temperature Display
SDP
Catalog No. 502875702 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO Series
KB Series
KBF PRO Series
KBF Series
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5702 KB Series
50-287-5634 KB PRO Series
50-287-5703 KBF PRO Series
50-287-5636 KBF Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-287-5702 Supplier BINDER™ Supplier No. 80122549
Only null left
Add to Cart
Add to Cart
Flexible Pt100 indoor temperature sensor
Platinum 100 ohm temperature sensor for sample temperature display or control.
Specifications
For Use With (Equipment) KB Series
Material Steel
Type Temperature Sensor