Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zhejiang Gongdong Medical Tech CO Ltd Specimen Tube with Red Cap, 10 mL

Catalog No. 12800100 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-800-100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-800-100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-Sterile Transport Tubes with separate red screw cap feature optically clear polypropylene (PP) for the collection and transportation of liquid specimens.

  • Optically Clear
  • Non-sterile
  • Red Screwcap Closure Included

Specifications

Closure Color Red
Closure Material Polypropylene
Color Clear
Capacity (Metric) 10 mL
Material Polypropylene
For Use With Liquid Storage and Transportation
Packaging Tubes: 800/Bag (2 Bags/Case), Caps: 200/Bag (8 Bags/Case)
Quantity 1600/Cs.
Sterility Non-sterile
Closure Type Screw Cap
Product Type Specimen Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.