Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin Dihydrochloride Pentahydrate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139810
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139810 25 g
S139767 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139810 Supplier G-Biosciences Supplier No. RC1102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spectinomycin is an aminocyclitol antibiotic isolated from Streptomyces spectabilis. It is effective against gram-positive and gram-negative bacteria, but most commonly used against Gonococcus species.

It has also been used to monitor cell fate during leaf development and as a selection marker in plants to display successful transformation of the Spcr gene. Spectinomycin functions by binding the 30s ribosomal subunit, inhibiting tRNA translation and protein synthesis. Resistance to this antibiotic is conferred through the aadA gene and mutated rpsE gene.

Chemical Identifiers

CAS 22189-32-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 25 g
Percent Purity 98%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.