Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin HCI, USP Grade, G-Biosciences
SCI_ED

Catalog No. S162503
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162503 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S162503 Supplier G-Biosciences Supplier No. RC868
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spectinomycin is an aminocyclitol antibiotic which acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis.

Its activity is generally modest, particularly against gram-positive bacteria. Some gram-negative bacteria are sensitive. Resistance in vitro may develop by chromosomal mutation or may be plasmid mediated.

Chemical Identifiers

CAS 22189-32-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 250 g
Percent Purity 98%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.