Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
SDP

Catalog No. 08607026 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
1000 kDa
20 kDa
3.5 to 5 kDa
300 kDa
50 kDa
8 to 10 kDa
Color Code:
Amber
Black
Blue
Green
Orange
Pink
Red
Violet
Yellow
Length (Metric):
50 mm
100 mm
160 mm
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Color Code Length (Metric)
08-607-026 0.5 to 1 kDa Orange 160 mm
08-607-012 100 kDa Blue 50 mm
08-607-023 3.5 to 5 kDa Black 50 mm
08-607-010 50 kDa Violet 160 mm
08-607-006 100 kDa Blue 100 mm
08-607-027 300 kDa Amber 50 mm
08-607-022 0.1 to 0.5 kDa Green 50 mm
08-607-021 8 to 10 kDa Yellow 50 mm
08-607-016 0.1 to 0.5 kDa Green 160 mm
08-607-031 1000 kDa Pink 100 mm
08-607-005 50 kDa Violet 50 mm
08-607-015 8 to 10 kDa Yellow 160 mm
08-607-032 1000 kDa Pink 160 mm
08-607-001 0.1 to 0.5 kDa Green 100 mm
08-607-028 300 kDa Amber 100 mm
08-607-008 50 kDa Violet 100 mm
08-607-025 0.5 to 1 kDa Orange 100 mm
08-607-030 1000 kDa Pink 50 mm
08-607-029 300 kDa Amber 160 mm
08-607-024 0.5 to 1 kDa Orange 50 mm
08-607-007 20 kDa Red 100 mm
08-607-020 3.5 to 5 kDa Black 100 mm
08-607-009 20 kDa Red 160 mm
08-607-000 8 to 10 kDa Yellow 100 mm
08-607-017 3.5 to 5 kDa Black 160 mm
08-607-011 100 kDa Blue 160 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 08-607-026 Supplier Spectrum™ Supplier No. G235063
Add to Cart
Add to Cart

Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)

Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Superior handling and leak protection
  • Resealable screw cap and pipet for easy loading and total recovery
  • Highest membrane purity, no heavy metals and sulfides
  • Minimizes volume specific dilution
  • 9 MWCOs ranging from 100 to 1,000,000 Daltons

Specifications

Product Type Float-A-Lyzer G2 Dialysis Device
MWCO 0.5 to 1 kDa
Membrane Cellulose Ester
Diameter (English) Membrane 0.39 in.
Diameter (English)Top 0.9 in.
Diameter (Metric) Membrane 10 mm
Diameter (Metric)Top 23 mm
Length (English) 6.2 in.
Length (Metric) 160 mm
Closure Material Polypropylene
Closure Type Screw Cap
Color Code Orange
Kit Contents Disposable pipette for 5 and 10mL, flotation ring
Quantity 12 Pack
For Use With (Application) Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.