Learn More
Molecular Devices Spectramax Id5 Multi-Mode Microplate Reader
Supplier:  Molecular Devices ID5BLG
Encompass_Preferred
Spectramax Id5 Multi-Mode Microplate Reader With Bottom Luminescence Read Mode Monochromator-Based Spectral Absorbance And Monochromator-Based And Filter-Based Fluorescence Luminescence Fluorescence Polarization And Time-Resolved Fluorescence On-Board Touchscreen For Standalone Acquisition With User Access Control Nfc Personalization Softmax Pro Software (4 Perpetual Licenses) Integrated Shaking And Temperature Control Power Cable Lan Cable And Standard Filter Set Including Excitation Filters Trf 350NM FP Polarized Filter Vertical And Horizontal (400NM - 750NM ) And Emission Filters Trf 490 NM Trf 616 NM FP Polarized Filter Vertical And Horizontal (400NM - 750NM ) Compatible With 6- To -384 Well Plates Compatible With Scanlater Western Blot Membrane Holder Spectradrop Micro-Volume Microplate And 22 Mm Spectracuvette Adapter Plate (All Sold Separately) Including 12 Months Parts And Labor Warranty Comes With 12 Months Mdcare Phd Technical Support Plan
Catalog No. 50-000-14718
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.