Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-3 Recombinant Rabbit Monoclonal Antibody (14H1L14)

Catalog No. PI701938 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI701938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI701938 Supplier Invitrogen™ Supplier No. 701938
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Cat and Dog Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Spectrins are principle components of a cell's membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. The protein encoded by this gene (SPTBN2), is called spectrin beta non-erythrocytic 2 or beta-III spectrin. It is related to, but distinct from, the beta-II spectrin gene which is also known as spectrin beta non-erythrocytic 1 (SPTBN1). SPTBN2 regulates the glutamate signaling pathway by stabilizing the glutamate transporter EAAT4 at the surface of the plasma membrane. Mutations in this gene cause a form of spinocerebellar ataxia, SCA5, that is characterized by neurodegeneration, progressive locomotor incoordination, dysarthria, and uncoordinated eye movements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta-3
Applications Immunocytochemistry
Classification Recombinant Monoclonal
Clone 14H1L14
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Sptbn2
Gene Accession No. O15020
Gene Alias beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spectrin Beta; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin beta, non-erythrocytic 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; spinocerebellar ataxia 5 protein; Spnb3; SPNB-3; SPTBN2
Gene Symbols Sptbn2
Host Species Rabbit
Immunogen Peptides corresponding to Human SPTBN2 (aa 1334-1351, 1733-1750 ).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.