Learn More
MilliporeSigma™ Supelco™ Spectroline™ CM UV-Viewing Cabinets
Provide long- and shortwave irradiation for viewing TLC plates, and for UV fluorescence or absorption demonstrations
Supplier: MilliporeSigma™ Supelco™ Z1694471EA
Description
- Portable, lightweight aluminum minicabinets with 4 and 6 W removable lamps that may be used for hand-held applications
- Lamps provide long- and shortwave irradiation for viewing TLC plates, and for UV fluorescence or absorption demonstrations
- Cabinets have a flexible, contoured viewer for built-in UV-absorbing window
- Easy-access felt curtains, and removable bottoms for placement over transilluminators for greater intensity and contrast
- Meets OSHA requirements, CE Compliant, and UL Listed
Compliance
Meets OSHA Requirements, CE Compliant, and UL Listed
Specifications
UV | |
6 W | |
12 x 9 x 6.5 in. | |
Viewing TLC Plates, and UV Fluorescence or Absorption Demonstrations | |
E-Series UV Lamp (6 W), and European 2-Pin Plug (Adapter Sold Separately) | |
CM-26 |
230 V | |
365/254 nm | |
6 W, 230 V, 50 Hz | |
50 Hz | |
Aluminum | |
UV Viewing Cabinet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spectroline is a registered trademark of Spectronics, Corp.
Spot an opportunity for improvement?Share a Content Correction