Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectromelt A 10, MilliporeSigma™

Catalog No. M1107831000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1107831000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1107831000 Supplier MilliporeSigma Supplier No. 1.10783.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
Synonym lithium tetraborate, di-lithium tetraborate
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Color White
Physical Form Solid
Quantity 1 kg
Linear Formula Li2B4O7
UN Number NONH for all modes of transport
Solubility Information 50g/L (20°C)
Formula Weight 169.12
Concentration or Composition (by Analyte or Components) Li2B4O7
Odor odorless
Packaging Plastic Bottle
Appearance White solid
Density Bulk Density: 330 kg/m3
Chemical Name or Material di-Lithium Tetraborate
Show More Show Less
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.