Learn More
United Scientific Diffraction Grating for SPECT01 Basic Spectrometer
Click to view available options
:
Diffraction Grating
Description
- Replacement diffraction grating with 600 lines per millimeter splits light into multiple beams for analysis in light and optics experiments
- 45 x 60mm (H x W) grating has a viewing area of 30 x 45mm (H x W)
- Dual glass plates protect grating from damage and provide visibility
- For use with the United Scientific SPECT01 spectrometer
Specifications
Specifications
| Type | Student Transmission Grating |
| For Use With (Equipment) | For SPECT01 Spectrometer. |
| Scale Type | Vernier |
| For Use With (Application) | To perform quantitative spectroscopy experiments, for student use in the classroom or lab |