Learn More
Interlight SPECTRONIC 20 HALOGEN
Supplier:  Interlight WW0DCR8
Encompass_Preferred
SPECTRONIC 20 HALOGEN: (for BAUSCH & LOMB) and others. Watts:17.88w, volts: 6.50, AMPS 2.75A
Catalog No. NC1874183
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.