Learn More
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer Accessories
Catalog No. 14380457
Change view
Click to view available options
Type:
Cell Holder
Cuvette Rack
Replacement Sample Compartment
SPECTRONIC Standards 2 Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-380-457 Cell Holder
14-385-495 Cuvette Rack
13-875-126 Replacement Sample Compartment
14-380-008 SPECTRONIC Standards 2 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-380-457 Supplier Thermo Scientific™ Supplier No. 840250500
Add to Cart
Add to Cart
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
The SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer ideal for routine applications. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.
Rugged, Practical Design
  • With no internal parts, there's nothing to wear out
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from acidic or oxidizing solutions
  • Navigation pad and on-screen menus are second nature to any modern user
  • Compact footprint saves space in your lab
  • Cuvette racks on both sides of the sample compartment help you keep track of samples
  • No computer or serial cable required; Just plug it in and you're ready to measure
The Perfect Tool for Routine Spectroscopic Measurements
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Standard sample compartment accommodates both 10mm square cuvettes and test tubes up to 25mm (1 inch) diameter
  • Room light immune optical system lets you measure tall test tubes with the lid open
  • Analyzer mode allows you to program standard methods
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
  • Available Thermo Scientific VISIONlite5 software, for your computer, controls the instrument and provides data manipulation, storage and report printing options
Not ready to part with your old Thermo Scientific Spec 20 or GENESYS 20
  • The SPECTRONIC 200 will emulate the interface and controls on your old instrument to integrate seamlessly into your laboratory
VISIONlite5 software — Volume pricing begins at eight copies and is available on request.
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Holder
For Use With (Equipment) SPECTRONIC 200/200E Spectrophotometers
Holds 100 mm Rectangular Cells
Shape Rectangular