Learn More
Spectroline™ Camera Adapters for UV Cabinets
SDP
Catalog No. FBCADS34M Shop All Spectronics Corporation Products
Change view
Click to view available options
4 x 5 in. field
8 x 10 in. field
Camera Adapter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
FB-CA-DS34M 4 x 5 in. field
FB-CA-35 Camera Adapter
FB-CA-DS34 8 x 10 in. field
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FB-CA-DS34M Supplier Spectroline™ Supplier No. CADS34/M
Snap-on adapters replace eyepiece on UV cabinets
  • Camera snaps onto adapter so that lens is perpendicular to plane of gel or TLC plate to give undistorted photograph of experimental results
  • Fit Spectronics Corporation Photodocumentation Camera