Description
- Improve contrast between the fluorescent area and the background by eliminating the “blue haze” interference
- Goggles (UVG–50) provide maximum protection from extended or high intensity UV exposure and adjust to fit the face
Compliance
Goggles meet ANSI specifications and OSHA standards.
Specifications
Specifications
| Product Type | Goggles |