Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectroline™ Replacement White Light Bulb
SDP

Catalog No. FBDLT10 Shop All Spectronics Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Lamps and Transilluminators
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
FB-DLT-10 Lamps and Transilluminators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB-DLT-10 Supplier Spectroline™ Supplier No. BLE1T230

missing translation for 'validMainframeMsg'

Fluorescent bulb for use with a variety of lamps and transilluminators

  • 8 Watts
  • 12 in. long

Specifications

Product Type Replacement Light Bulb
Color White
For Use With (Equipment) Lamps and Transilluminators
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.