Learn More
Spectrocell Inc Spectrophotometer Cells 100 mm, Rectangular
Supplier:  Spectrocell Inc R3100
Encompass
Open top quartz 100 mm path length FUV cell with PTFE lid and 2 polished windows. Most standard types supply with 1% matching. All cells and cuvettes are available with anti-reflection (AR) coatings or mirror coatings for an additional charge. Top: Open top cell, Includes: PTFE lid, Path Length: 100 mm, Volume: 35.0 mL, Windows: 2 (polished), Spectral Range: 170-2200 nm, Type: 1
Catalog No. NC1775042
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.