Learn More
MilliporeSigma™ Supelco™ Spectrophotometer Cuvettes
Packaged in styrofoam boxes to assure cuvettes are dust- and scratch-free
Supplier: MilliporeSigma™ Supelco™ C5291100EA
Description
- These cuvettes are incompatible with some organic solvents
Specifications
Spectrophotometer Cuvette | |
≥2.5 mL | |
100/Pk. | |
340 to 800 nm |
Without Stopper | |
Polystyrene | |
10 mm | |
UV/Vis Spectroscopy |