Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ MediaKap™ and MediaKap Plus HF Filters
SDP

Catalog No. 086712B Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 L
10 to 25 L
2 to 5 L
25 to 50 L
5 to 10 L
Connectors:
0.25HB/0.25HB
Female Luer Lock/0.25HB
VarHB/VarHB
Includes:
Automatic vent
Automatic vent, filling bell
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connectors Includes
08-671-2B 0.2 to 2 L Female Luer Lock/0.25HB Automatic vent, filling bell
08-607-033 0.2 to 2 L Female Luer Lock/0.25HB Automatic vent
08-671-2E 5 to 10 L 0.25HB/0.25HB Automatic vent
08-671-2G 10 to 25 L VarHB/VarHB Automatic vent, filling bell
08-607-039 25 to 50 L VarHB/VarHB Automatic vent, filling bell
08-671-4C 2 to 5 L Female Luer Lock/0.25HB Automatic vent
08-671-4F 10 to 25 L VarHB/VarHB Automatic vent, filling bell
08-671-4E 5 to 10 L 0.25HB/0.25HB Automatic vent, filling bell
08-671-4D 2 to 5 L Female Luer Lock/0.25HB Automatic vent, filling bell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 08-671-2B Supplier Spectrum™ Supplier No. ME2M02B12S

0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium

Specifications

Connectors Female Luer Lock/0.25HB
Filter Type MediaKap-2
Material Polycarbonate, Polyurethane
Material (Membrane) MCE
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 35 cm2
Flow Rate 400 mL/min. at 10psi
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Includes Automatic vent, filling bell
No. per Case 12 Case
Type Bottle Top Filter
Volume (Metric) 0.2 to 2 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.