Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
SDP

Catalog No. 08670264 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
24 mm
MWCO:
0.1 to 0.5 kDa
0.5 kDa
1 kDa
100 kDa
25 kDa
5 kDa
50 kDa
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) MWCO
08-670-264 20 mm 25 kDa
08-671-25 20 mm 0.5 kDa
08-671-27 20 mm 1 kDa
08-671-24 24 mm 0.5 kDa
08-671-26 15 mm 1 kDa
08-671-30 15 mm 5 kDa
08-671-31 20 mm 5 kDa
08-671-34 6.4 mm 50 kDa
08-671-37 20 mm 50 kDa
08-671-39 10 mm 100 kDa
08-671-41 20 mm 100 kDa
08-750-10B 6.4 mm 25 kDa
08-750-2D 10 mm 0.1 to 0.5 kDa
08-750-6B 6.4 mm 5 kDa
08-801-27 15 mm 25 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 08-670-264 Supplier Spectrum™ Supplier No. 131348
Add to Cart
Add to Cart

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Product Type Spectra/Por Dialysis Membrane Tubing
MWCO 25 kDa
Material Cellulose Ester
Diameter (Metric) 20 mm
Diameter (English) 0.78 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Supplied in 0.1% sodium azide solution
Temperature Range (English) 39.2°F to 98.6°F
Temperature Range (Metric) 4°C to 37°C
Quantity 33 Pack
For Use With (Application) Ideal for desalting, purification, and molecular separation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.